Orders LinkContactUsLinkComponent
BoneClones® Replica Teeth
BoneClones® Replica Teeth
Certificates
BoneClones® Replica Teeth
BoneClones® Replica Teeth
4 Options Available Below
Product Details & Documents
Compare tooth shape based on diet.
  • Explore the influences of diet and use in tooth shape
  • Highly detailed replicas from real specimens
Teeth serve many different functions in the animal world, each of the teeth below have evolved based on carnivorous nature or the animal. What does the structure of each tell about the feeding habits of the animal?
Product Family Options
Product Information
  • No. of Parts
    Length
    Availability
    Price
  • 470095-170
    Great white shark tooth
    Great white shark tooth
    1
    6.25 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $426.41
    $426.41
    Each (1UN)
       $426.41
    Supplier Number: KO-223
  • 470095-200
    Orca tooth
    Orca tooth
    1
    6.25 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $692.89
    $692.89
    Each (1UN)
       $692.89
    Supplier Number: KO-012
  • 470095-132
    Polar bear tooth (Molar)
    Polar bear tooth (Molar)
    1
    3.75 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $586.29
    $586.29
    Each (1UN)
       $586.29
    Supplier Number: KO-013
  • 470312-206
    Hippo tooth
    Hippo tooth
    1
    5 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $558.00
    $558.00
    Each (1UN)
       $558.00
    Supplier Number: KO-214