Orders LinkContactUsLinkComponent
ScoutPlus Binoculars
ScoutPlus Binoculars
ScoutPlus Binoculars
Catalog # 470227-902
Supplier:   CARSON OPTICAL
Número de proveedor #:  JD-025
ScoutPlus Binoculars
Catalog # 470227-902
Supplier:  CARSON OPTICAL
ScoutPlus Binoculars
ScoutPlus Binoculars
ScoutPlus Binoculars
Price
$1,508.41
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
A 10X binocular with superior optics and a compact size.
  • Fully coated lenses
  • 9.9 oz. weight
Ordering information: Includes pouch, neck strap, and lens cloth.
Especificaciones
  • Magnification:
    10×
  • Description:
    ScoutPlus Binoculars