MicroPen Microscope
MicroPen Microscope
Catalog # 470227-898
Supplier: CARSON OPTICAL
Número de proveedor #: MP-300
Jump to:
Product Details & Documents
A microscope the size of a pen! Use this in an industrial setting, the classroom, or the back yard.
- 24X–53X magnification
- 3G3 button cell batteries
- Industrial use
- Classroom use
- Built-in LED
- Focus and zoom adjustments
Especificaciones
- Magnification:24× - 53×
- Description:MicroPen Microscope