MagniVisor Deluxe
Catalog # 470227-880
Supplier: CARSON OPTICAL
Specifications
- Aumento:1.5×, 2×, 2.5×, 3×
- Descripción:MagniVisor Deluxe
- Referencia:470227-880
Specifications
About this item
A hands free magnifier with adjustable headband, 4 lenses and removable LED lamp.
- 4" lens diameter
- 3 AAA batteries
- 5.8 oz. weight