Orders LinkContactUsLinkComponent
MicroBrite Plus Microscope
MicroBrite Plus Microscope
MicroBrite Plus Microscope
Catalog # 470227-866
Supplier:   CARSON OPTICAL
Supplier #:  MM-300
MicroBrite Plus Microscope
Catalog # 470227-866
Supplier:  CARSON OPTICAL
MicroBrite Plus Microscope
MicroBrite Plus Microscope
MicroBrite Plus Microscope
Price
$445.07
Log in for availability
Each (1UN)
Product Details & Documents
A 60-120X microscope that is lightweight and portable.
  • 60X–120X magnification
  • Lightweight
  • Fits easily in a pocket
  • Built-in LED for a bright, clear image
Specifications
  • Magnification:
    60× - 120×
  • Description:
    MicroBrite Plus Microscope