Orders LinkContactUsLinkComponent
Test Sieve Cleaning Brushes, W.S.Tyler
Test Sieve Cleaning Brushes, W.S.Tyler
Supplier:   W.S. Tyler
Test Sieve Cleaning Brushes, W.S.Tyler
Supplier:  W.S. Tyler
Certificates
Test Sieve Cleaning Brushes, W.S.Tyler
Test Sieve Cleaning Brushes, W.S.Tyler
2 Options Available Below
Product Details & Documents
These brushes are used for cleaning test sieves.
Product Family Options
Información del producto
  • Bristle Material
    Handle Material
    Availability
    Price
  • 80086-446
    Brass Test Sieve Cleaning Brush for Cleaning Sieves with 100 Mesh and Coarser
    Brass Test Sieve Cleaning Brush for Cleaning Sieves with 100 Mesh and Coarser
    Brass
    Wood
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,220.93
    $1,220.93
    Each (1UN)
       $1,220.93
    Supplier Number: 8576
  • 80086-444
    Nylon Test Sieve Cleaning Brush for Cleaning Sieves with 120 Mesh and Finer
    Nylon Test Sieve Cleaning Brush for Cleaning Sieves with 120 Mesh and Finer
    Nylon
    Wood
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,314.71
    $1,314.71
    Each (1UN)
       $1,314.71
    Supplier Number: 8577