Digital Stroboscope
Product Details & Documents
Freeze time in the apparent motion of a vibrating string.
- Unit has setting in mSec, Hz and rpms
Digital stroboscope can appear to slow down fast periodic motions for a bystander. This is why stroboscopes are great for the physics classroom or calibrating revolutions from a device such as a motor. The intense xenon lamp emits a burst of light which can be synchronized with a particular target (sometimes to make something appear still, or to take a frequency measurement, etc.). This high quality unit uses an internal microcontroller as the processing unit with built in display chip. The controller drives a 7 segment LED to present real-time flashing frequency data. One of the most popular demonstrations to do with this device is the standing wave string.
Especificaciones
- Description:Digital Stroboscope