Magnet Probe
Magnet Probe
470227-642
: PH0798MP
Visualize the shape of the magnetic field around a magnet.
- Strong plastic construction
Consists of a small 19 mm length alnico bar magnet mounted in freely rotating gimbal. It rotates in 3D so that all x, y and z co-ordinates, three-dimensional magnetic force fields, can be mapped.
- Description:Magnet Probe