Vacuum Gauge
Vacuum Gauge
470227-544
: HVG-004
This bourdon style vacuum gauge features a 2" Dial with a range of 30-0" Hg.
- Accuracy +-3%
- Operating Temperature range -40 to 150°F
The connection size is 1/4" NPT and is located on the bottom of the gauge for easy mounting.
- Description:Vacuum Gauge