Orders LinkContactUsLinkComponent
HPLC Columns, YMC-Pack ODS-AM
HPLC Columns, YMC-Pack ODS-AM
Analytical, ODS-AM, 120 Å, 5 µm, 4.60 mm, 150 mm
Catalog # 97028-794
Supplier:   YMC AMERICA, INC
Número de proveedor #:  AM12S05-1546WT
HPLC Columns, YMC-Pack ODS-AM
Catalog # 97028-794
Supplier:  YMC AMERICA, INC
HPLC Columns, YMC-Pack ODS-AM
Analytical, ODS-AM, 120 Å, 5 µm, 4.60 mm, 150 mm
Price
$42,454.04
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
ODS-AM is a high quality analytical C18 material, with a very tight specification.
  • Long term reproducibility
  • Highly versatile
  • Ideal choice for establishing validated HPLC analysis
ODS-AM has appropriate selectivity for polar to moderately nonpolar pharmaceuticals, organic intermediates, biological and natural products found in the chemical and pharmaceutical industry.
Especificaciones
  • Column Format:
    Analytical
  • Ø int.:
    4.60 mm
  • Particle Size:
    5 µm
  • Pore size:
    120 Å
  • Length:
    150 mm
  • Phase:
    ODS-AM
  • Carbon load:
    17%
  • Cat. No.:
    97028-794
  • Description:
    97028-794
  • Endcapped:
    Yes
  • pH range:
    2 - 7.5
  • Surface area:
    330 m2/g