Orders LinkContactUsLinkComponent
VertiKlean® MAX™ Mop Wipe System
VertiKlean® MAX™ Mop Wipe System
Supplier:  CONTEC, INC. MX
Certificates
VertiKlean® MAX™ Mop Wipe System
VertiKlean® MAX™ Mop Wipe System
5 Options Available Below
Product Details & Documents
The VertiKlean® MAX™ Mop Wipe System offers a quick and easy solution to mopping floors, walls, and other flat surfaces in any controlled environment. This versatile mop works with a variety of Contec® mop wipe covers, enabling the user to customize the mop to the cleaning task.
  • Stainless steel frame is chemical resistant
  • Autoclavable; can be sterilized by most common methods including gamma irradiation, ethylene oxide, or autoclave (recommended cycle is 121°C for 30 minutes)
  • Easy to attach and remove mop covers
  • Variety of fabric options available
  • Removes contaminants easily from the environment
  • Extends the life of the VertiKlean® MAX™ foam mop head
  • Presaturated wipe covers eliminate the need for a bucket and wringer
  • Slim profile allows easy access to confined spaces for efficient cleaning and disinfection
  • Thin, lightweight design makes it easy to clean floors, walls, and ceilings
Dry and presaturated single-use wipes fit over the VertiKlean® MAX™ foam mop head and QuickConnect frame to remove contamination such as powder, dirt, and dust from floors, walls, and ceilings. The VertiKlean® MAX™ Mop Wipe System provides excellent surface contact and conformity.
It can be used to apply disinfectant solutions to surfaces when used with a bucket and wringer. Most importantly, when used for disinfection, the mop wipe covers entrap microorganisms, enabling them to be removed from the environment with each change of wipe cover.
 : Presaturated or dry wipes are available
Product Family Options
Información del producto
  • Size
    Packed
    Availability
    Price
  • 76006-334
    Quiltec® Mop wipes, 100% polyester quilted
    Quiltec® Mop wipes, 100% polyester quilted
    25.4×43 cm (10×17")
    20/Bag, 12 Bags/Case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (240UN)
    Case (240UN) - $11,189.58
    $11,189.58
    Case (240UN)
       $11,189.58
    Supplier Number: LWQT0100
  • 10816-268
    QuickConnect™ Mop wipe frame, stainless steel
    QuickConnect™ Mop wipe frame, stainless steel
    29×4.4×10.8 cm (11.5×1.75×4.25")
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,922.20
    $4,922.20
    Each (1UN)
       $4,922.20
    Supplier Number: 2652K
  • 76006-338
    Amplitude™ Sigma™ Mop wipes, nonwoven cellulose/polyester
    Amplitude™ Sigma™ Mop wipes, nonwoven cellulose/polyester
    30.5×40.6 cm (12×16")
    50/Bag, 20 Bags/Case
    Discontinued
    Supplier Number: -
  • 10119-378
    Medium VertiKlean® MAX™ Sterile ribbed microfiber mop head
    Medium VertiKlean® MAX™ Sterile ribbed microfiber mop head
    33×10.2×2.5 cm (13×4×1")
    1/Bag, 28 Bags/Case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (28UN)
    Case (28UN) - $16,056.36
    $16,056.36
    Log in Para disponibilidad
    Each (28UN)
    Each (28UN) - $16,056.36
    $16,056.36
    Case (28UN)
       $16,056.36
    Each (28UN)
       $16,056.36
    Supplier Number: VKMX0275
  • 89495-244
    Medium VertiKlean® MAX™ Ribbed microfiber mop head
    Medium VertiKlean® MAX™ Ribbed microfiber mop head
    33×10.2×2.5 cm (13×4×1")
    4/Bag, 7 Bags/Case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (28UN)
    Case (28UN) - $9,612.66
    $9,612.66
    Case (28UN)
       $9,612.66
    Supplier Number: VKMX0225
  • 10171-158
    Medium VertiKlean® MAX™ Sealed edge mop head
    Medium VertiKlean® MAX™ Sealed edge mop head
    38×12.7×1 cm (15×5×0.375")
    4/Bag, 4 Bags/Case
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (16UN)
    Case (16UN) - $6,206.91
    $6,206.91
    Case (16UN)
       $6,206.91
    Supplier Number: VKSE0200