Avantor® Apex and Apex ll, HPLC Columns
75798-984
: Avantor
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:5 µm
- Tamaño de poro:100 Å
- Descripción:Apex ODS column
- Longitud:250 mm
- Referencia:75798-984
- Packing Material:Silica
- Phase:ODS
- Nº de proveedor:4M25310
Avantor® Apex and Apex ll phases are recommended for routine analyses.
- Conventional 100 Å pore size spherical silica
- Narrow particle distribution
- Controlled surface area