Orders LinkContactUsLinkComponent
Aquaponics System
Aquaponics System
Aquaponics System
Certificates
Aquaponics System
Aquaponics System
2 Options Available Below
Product Details & Documents
Convienent Classroom Farming.
  • All acrylic construction
  • Super easy set up
  • Adjustable water flow to plant trays
  • Includes plastic net pots, growing medium, and pump
Classroom Aquaponics engages students in ecology, biology, earth systems, math, history and literature while pairing fish and plants in one integrated system. Every Classroom Aquaponics unit is equipped with an instructional manual. In addition to growing vegetables in a sustainable process, educators instruct on germination rates, fish anatomy and the nitrogen cycle.
Ordering information: Unfortunately due to the nature of the product, this item is not returnable.
Product Family Options
Product Information
  • Availability
    Price
  • 470227-226
    Aquaponics System
    Aquaponics System
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $23,769.48
    $23,769.48
    Each (1UN)
       $23,769.48
    Supplier Number: 91000
  • 470231-364
    Aquaponics Kit with Activities
    Aquaponics Kit with Activities
    Log in for availability
    Log in for availability
    Kit de 1
    Kit de 1 - $24,340.86
    $24,340.86
    Kit de 1
       $24,340.86
    Supplier Number: 635-9100