DURX® 570 Polyester/Cellulose Nonwoven Wipers, Berkshire
Catalog # 76003-806
Supplier: BERKSHIRE CORP
Specifications
- Anchura:22.9 cm (9")
- Descripción:Knife-Cut Edge
- Longitud:22.9 cm (9")
- Matriz:Nonwoven Polyester/Cellulose
- Referencia:76003-806
- Empaquetado:Stacked
Specifications
About this item
Ideal for cleaning work surfaces and spill pickup, general-purpose wiping, and polishing of surface areas, including optics.
- Smooth, highly absorbent, and durable
- Low extractable levels and particle counts
- Chemically compatible with common cleaning and disinfecting solutions
- Autoclavable
- Economical
Durx® 570 Wipers are free of chemical additives, and designed for use in ISO Class 5 – 8 controlled environments. Specifically recommended for use in ISO Class 5 – 8 (Class 100–100,000) cleanroom applications.