Fiber Optics Voice and Data Kit
Fiber Optics Voice and Data Kit
Catalog # 470227-036
Supplier: ELENCO ELECTRONICS INC SE
Número de proveedor #: FO30K
Jump to:
Product Details & Documents
Learn the basics of fiber optics communications while you build this exciting kit.
- Includes instruction manual
- Includes 3 foot fiber optic cable
Transmit your voice or a 1kHz signal through the fiber optic cable to a fiber optic receiver. Comes complete with all of the necessary components; including PC boards; microphone; speaker and fiber optic cable. Transmitter and receiver (BOTH included) on separate boards; can be located hundreds of feet apart. Use for science projects.
Documents
Manual 470227-036
Manual
Specifications
- Description:Fiber Optics Voice and Data Kit