Accessories for Reaction Vessels, Chemglass
80061-174
Small Business Enterprise
- Descripción:O-Ring, Silicone
- Referencia:80061-174
- Para ser utilizado con:150 mm Flange
- Nº de proveedor:CG-147-33
Flange is formed with a groove in the flat portion of the mating surface to accept an O-Ring.
It also has a ground surface, permitting assembly with either the CG-148 flat flange or CG-149 lid blank using grease.
In the greased application, the O-Ring groove prevents grease from entering the reaction kettle and causing possible contamination.
Outer taper of flange is formed at a 45° angle to accept the CG-141 clamp.