Plastic Coverslips
Catalog # 470227-310
Supplier: RAYLABCON INC (NY)
Specifications
- Descripción:Plastic cover slips
- l×a:22×22 mm
- Grosor:0.25 mm
- Referencia:470227-310
Specifications
About this item
Efficient, disposable slides are manufactured from optically clear vinyl.
- Made of clear vinyl plastic
- Separated from each other by tissue paper; no more frustrating plastic covering to remove
- Unbreakable and safer than glass
- Used in bioloigical and medical labs, as well as for educational applications
Slides will not break, scratch, or cut fingers. Ideal for wet mounts.
Order by the each (100 cover slips), or by the pack (10 packs of 100 cover slips).