VWR® Chemical Splash Goggles
Catalog # 75982-628
Supplier: VWR International
Specifications
- Descripción:Chemical splash goggle
- Referencia:75982-628
- Recubrimiento de lentes:Anti-fog, anti-scratch
- Material del marco:PVC
- Lens Material:PC
Specifications
About this item
Wide vision goggles provide high levels of comfort and maximum splash protection.
- Wraparound modern style provides excellent panoramic vision
- Fits most facial contours
- Comfortably fits over most prescription glasses
- Indirect ventilation
- Provides 99.9% UV protection
These high-quality safety goggles feature a modern, wraparound style with fogless, distortion-free panoramic lenses. They are equipped with indirect venting for liquid splash protection and internal relief slots to reduce interference with prescription glasses. The wide elastic strap with adjustable buckle attaches to the frame with a swivel clip for a comfortable fit. The nose area is split for greater comfort and the wide facial flange provides a secure seal to almost every face.
Certifications: Meet ANSI Z87+ 1-2015 standards.