Cleaning Brushes, Burkle
Catalog # 75997-618
Supplier: BURKLE INC
Specifications
- Bristle Material:PVC
- Cabeza del cepillo Ø:25 mm (0.98")
- Handle Material:Stainless Steel
- Descripción:Cleaning Brushes
- Largo total:100 cm (39.37")
- Referencia:75997-618
Specifications
About this item
Cleaning brushes for thorough cleaning.
PVC/stainless steel
Packaging: One brush per package.