VWR® Certified Reference Materials, Oxide
2 kg, Gold, 0 - 10 / 100 - 1000 ppm, Au/Ag, Oxide, Jar
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
: 21002131
VWR® Certified Reference Materials, Oxide
Catalog # CA76202-564
Supplier: KLEN INTERNATIONAL (74) PTY LTD.
CAS Number:
Jump to:
Product Details & Documents
KLEN's CRMs are certified products containing a range of analytes including Au, Ag, Pt, and Pd at various ppm levels
: Other sizes (30g, 50g and 150g sachets) are available, please contact your Avantor sales representative for more information.
Especificaciones
Especificaciones
- Concentration:0 - 10 / 100 - 1000 ppm
- Matrix:Oxide
- Pack type:Jar
- Size:2 kg
- Analyte:Au/Ag
- Element:Gold