Leica EM TP Automatic Tissue Processor, Leica Microsystems
Catalog #
Jump to:
Product Details & Documents
Automated routine tissue processor for EM and LM sample preparation.
- Automated tissue processing
- Closed processing chamber
- High throughput
The innovative Leica EM TP is the first tissue processor designed for EM and LM resin processing that features a heating/cooling system with pre-heat and pre-cool capabilities to maintain a constant processing temperature.
It includes an easy-to-operate membrane control panel. The sample carousel holds 24 EM or 12 LM vials and utilizes various specialized baskets and capsules. An exhaust system supports safer use of toxic substances.
Caution: For research use only.
Specifications
- Cat. No.:S1174-500MG
- M-Clarity quality level:MQ100
- Storage conditions:RT
- MDL No.:MFCD00006626
- Melting point:185 - 187 °C (lit.)
- Synonym(s):α-ᴅ-Glc-(1→2)-β-ᴅ-Fru; α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside; β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside; ᴅ(+)-Saccharose; Sugar
- Molecular weight:342,30
- CAS No.:57-50-1
- Shipping conditions:STD