PCR Tubes, Watson Bio Lab
Individual PCR tubes, Natural, 0.2 ml, Dome-shaped cap
Catalog # 75982-954
Supplier: WATSON BIO LAB USA
Supplier #: 137-212C
Jump to:
Product Details & Documents
The side walls are uniformly thin, which enables PCR with superior heat exchange.
- Thin wall, high transparency
- Nuclease-free certified
Flat and dome caps are available.
All tubes feature autoclavable polypropylene construction.
The flat top, smooth and superior in optical transparency, is the most suitable for Real Time PCR. They also offer writing space.
Specifications
- Cap Type:Dome-shaped cap
- Capacity:0.2 ml
- Color:Natural
- Description:Individual PCR tubes