Round-Bottom Standard Microtubes, Watson Bio Lab
Round-Bottom Standard Microtube, 0.6 ml
Catalog # 75982-888
Supplier: WATSON BIO LAB USA
Número de proveedor #: 130-806C
Jump to:
Product Details & Documents
These user-friendly standard microtubes feature an easy-opening, flat-top cap.
- Flat Cap
- High Transparency
- RNase-/DNase-/Human DNA-free
- Autoclavable Polypropylene Construction
- Scale Graduations
The flat cap provides a space to write. Users can autoclave the polypropylene tube. Graduated scale makes measuring simple.
Accessories information: Pestles, storage racks, and cap locks are available separately.
Especificaciones
- Volume:0.6 ml
- Description:Round-Bottom Standard Microtube