Pipette Tips, Watson Bio Lab
Graduated, Filtered, 1000 µl, +, +, Long
Catalog # 75982-816
Supplier: WATSON BIO LAB USA
Supplier #: 124P-1000S
Jump to:
Product Details & Documents
Watson offers pipette tips in a wide variety of volumes, lengths, and widths.
- Thin-walled tips available
- Soft-fitting tips available
- Tips come in short, standard, long, and extra-long sizes
- Large bore tips available
- Graduated tips provide simple measurement
Accessories information: Empty racks are available separately.
Especificaciones
- Volume:1000 µl
- Length:Long
- Filter:+
- Sterile:+
- Tip Style:Graduated, Filtered