Ward's Science Essentials® Basic Minerals Collection
Basic Minerals Collection, 1 - 1½", 12
470226-468
: Avantor
Introduce your students to a broad selection of minerals.
- Contains twelve specimens and photo key
- Perfect for teaching basic properties of minerals
Ward’s Science Essentials
Engage and inspire students to explore in-demand STEM careers with Ward’s Science Essentials. These multidisciplinary activities kits allow you to easily build STEM concepts into every lesson – with plenty of aha moments along the way.
Experience
Create real-world scenarios in the classroom, helping students see the role of STEM in everyday life.
Collaborate
Group-driven activities and challenges foster teamwork and build leadership, communication, and problem-solving skills.
: Collection contains Apatite, hornblende, muscovite, hematite, limonite, milky quartz, calcite, graphite, pyrite, halite, selenite and microcline.
- Description:Basic Minerals Collection
- No. of Samples:12
- Specimen Dimensions:1 - 1½"