Orders LinkContactUsLinkComponent
NeutraKlean Detergent, Contec®
NeutraKlean Detergent, Contec®
NeutraKlean Detergent, Contec®
Supplier:  CONTEC, INC. MX
Certificates
NeutraKlean Detergent, Contec®
NeutraKlean Detergent, Contec®
2 Options Available Below
Product Details & Documents
Contec® NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms.
  • Gentle and low-foaming detergent is ideal for low level cleaning in life science cleanrooms
  • Neutral pH makes it suitable for use in all areas of cleanroom
  • Filtered to 0.2 microns in a ISO Class 7 cleanroom ensures the product is free from contamination and particulates
  • Irradiated at no less than 25 kGy and provided with lot specific sterility certification
  • Low level of surfactant provides low residue which can be easily removed
  • Trigger spray and "bag in bottle" protected system prevents bottle contents from becoming contaminated during use
With a pH after irradiation between 6.5 and 7.5, NeutraKlean is ideal for carrying out the low-level cleaning required after maintenance, and to remove disinfectant residues and general soil. The use of a surfactant/detergent helps to break the binding of particles and dirt to a surface and allows them to be more easily removed.
Ready to use, the product is available in a 1 L trigger spray with a “bag in bottle” system to protect the contents during use. Both the sterile and filtered products are provided double bagged to aid the transfer of the product into a pharmaceutical cleanroom, isolators, and RABS. The linear tear bags are easy to open, even when wearing gloves.
Product Family Options
Product Information
  • Size
    Packaging
    Availability
    Price
  • 75982-114
    Irradiated NeutraKlean solution, neutral detergent with purified water
    Irradiated NeutraKlean solution, neutral detergent with purified water
    1 L
    Trigger spray bottle
    Log in for availability
    Log in for availability
    Case (6UN)
    Case (6UN) - $6,166.61
    $6,166.61
    Case (6UN)
       $6,166.61
    Supplier Number: SBT103NK
  • 75982-116
    Irradiated NeutraKlean solution, neutral detergent with purified water
    Irradiated NeutraKlean solution, neutral detergent with purified water
    5 L
    Bottle, capped
    Log in for availability
    Log in for availability
    Case (2UN)
    Case (2UN) - $4,226.95
    $4,226.95
    Case (2UN)
       $4,226.95
    Supplier Number: FBC503NK