Miscellaneous Dissolution Accessories, Quality Lab Accessories
Ever-Clear Dissolution Water Bath Preservative, Single Dose, 12 pack
Catalog # 75856-092
Supplier: QUALITY LAB ACCESSORIES
Supplier #: EVERCLEAR-12
Miscellaneous Dissolution Accessories, Quality Lab Accessories
Catalog # 75856-092
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
Everclear Water Bath Preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths.Use with DEIONIZED water
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, Rotating Bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE Suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.
Especificaciones
- Description:Ever-Clear Dissolution Water Bath Preservative, Single Dose, 12 pack