Orders LinkContactUsLinkComponent
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Certificates
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
1 Options Available Below
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Product Family Options
Información del producto
  • Availability
    Price
  • 11010-218
    Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Discontinued
    Supplier Number: DISASY-06
  • 11010-222
    Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Discontinued
    Supplier Number: DISTUB-06
  • 11010-360
    Clear Glass Disintegration Beaker with Flat Top, 900 mL
    Clear Glass Disintegration Beaker with Flat Top, 900 mL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $3,997.15
    $3,997.15
    Each (1UN)
       $3,997.15
    Supplier Number: GLA905-DB