Knife Switches
Supplier: VWR International
Jump to:
Acerca de este artículo
Bigger design for easy connections.
- Easy-to-connect
- Unbreakable
- Teaching tool for open circuit lessons
- Rated at 5A
- Four configurations available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits
4 Options Available Below
Product Details & Documents
Bigger design for easy connections.
- Easy-to-connect
- Unbreakable
- Teaching tool for open circuit lessons
- Rated at 5A
- Four configurations available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits
Product Family Options
Información del producto
- AvailabilityPrice
- 470225-202Knife Switch Single Pole, Single ThrowKnife Switch Single Pole, Single ThrowEach (1UN)$47.83Case de 50 (1UN)$52.13
- 470225-204Knife Switch Single Pole, Double ThrowKnife Switch Single Pole, Double ThrowEach (1UN)$1.80Case de 50 (1UN)$90.00
- 470225-206Knife Switch Double Pole, Single ThrowKnife Switch Double Pole, Single ThrowEach (1UN)$2.15Case de 50 (1UN)$107.50
- 470225-208Knife Switch Double Pole, Double ThrowKnife Switch Double Pole, Double ThrowEach (1UN)$2.60Case de 50 (1UN)$130.00