Making Dew Kit
Making Dew Kit, 15
Catalog # 470225-386
Supplier: AMERICAN SCIENTIFIC LLC SE
Número de proveedor #: 9447-15
Jump to:
Product Details & Documents
Get hands on with dew and condensation.
- Includes teacher guide
- Thermometers included
This kit help students understand how dew and frost are formed. They will also understand concepts of condensation, dew point, evaporation, humidity, and sublimation. Making Dew classroom pack contains materials for 15 pairs of students.
Ordering information: Includes 15 metal cans, 15 metal backed thermometers, and a Teacher's Guide.
Especificaciones
- Description:Making Dew Kit
- No. of Lab Groups:15