Orders LinkContactUsLinkComponent
Flask Brushes, Justman Brush
Flask Brushes, Justman Brush
Flask Brushes, Justman Brush
Certificates
Flask Brushes, Justman Brush
2 Options Available Below
Product Details & Documents
These brushes with special construction puts the cleaning action directly where it is needed.
The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.
Product Family Options
Información del producto
  • Bristle Color
    Brush head Ø
    Handle Material
    Availability
    Price
  • 17145-050
    For 25 mL flasks
    For 25 mL flasks
    Black
    44 mm (1 3/4")
    Wire
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $200.36
    $200.36
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $218.38
    $218.38
    Each (1UN)
       $200.36
    Paquete de 12 (1UN)
       $218.38
    Supplier Number: J620368
  • 17145-052
    For 50 mL flasks
    For 50 mL flasks
    Black
    44 - 54 mm
    Wire
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $135.80
    $135.80
    Log in Para disponibilidad
    Paquete de 12 (1UN)
    Paquete de 12 (1UN) - $1,629.60
    $1,629.60
    Each (1UN)
       $135.80
    Paquete de 12 (1UN)
       $1,629.60
    Supplier Number: J620370