DNA LoBind Tubes, Eppendorf
Product Details & Documents
DNA LoBind tubes improve recovery of nucleic acids by reducing their absorption to the tube wall.
- LoBind material guarantees maximum sample recovery
- Free of surface coating
- Precise lid sealing for minimal evaporation rates
The LoBind material guarantees maximum sample recovery for improved assay results. Free of surface coating (e.g., silicone) to minimize the risk of sample interference. Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors. Rated up to 30000×g centrifugation speed for molecular biology applications. Use for preparation or storage of DNA and RNA samples, forensic trace analysis, preparing dilution series in quantitative qPCR, sample preparation in next-generation sequencing, and creation of genomic or oligonucleotide libraries.
A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100% recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination.
Documents
Eppendorf Centrifuge Tubes
Product Literature
Especificaciones
- Volume:50 ml
- Description:DNA LoBind tubes, colorless
- Packaging:—
- Bottom Style:Conical bottom