Acerca de este artículo
This sterilizer offers fully automatic sterilization from start to finish with high-pressure steam sterilization and drying steps.
- Automatic high pressure steam sterilization system
- Drying material can be set according to sample material, quantity, and more
- Digital, interactive key-input system
- Quick sample drying capability
Operations can be carried out with an interactive key-input system, and a quick sample drying capability makes samples ready to use right after sterilization
The drying temperature can be set according to sample material, quality, and more. Self-diagnostic functions make operation safe and error recovery fast, and a condensation collector safely neutralizes high-temperature exhaust steam.
The front panel level indicator can quickly confirm the drain bottle water level, and the drain valve is on the front for easy access. The absence of protrusions in the sterilization chamber makes insertion and removal of baskets and other items simple. The sterilizer features a stainless steel interior.
Especificaciones
- Catalog No:
- 75846-584
- 75846-586
- Tipo de control:
- Digital PID Controller
- Digital PID Controller
- Temporizador:
- 1 Minute to 99 Hours and 59 Minutes, 100 to 999 Hours/1 Minute or 1 Hour
- 1 Minute to 99 Hours and 59 Minutes, 100 to 999 Hours/1 Minute or 1 Hour