SatPax® 550 Pre-Wetted 100% Meltblown Polypropylene Cleanroom Wiper, Berkshire
Proveedor: BERKSHIRE CORP
Certificados
Acerca de este artículo
SatPax® 550 wipers are exceptionally clean. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper.
- 100% meltblown polypropylene
- Pre-wetted with 70% IPA/30% DI water
- Re-sealable solvent resistant packaging
- Knife cut edges
- Excellent wet strength and abrasion resistance
- Reduces alcohol usage and preparation/handling cost
- Increases cleaning efficiency
SatPax® 550 wipers feature very low extractables and ions, as well as low particles and fibre counts.
Designed for use in ISO Class 5 and higher cleanroom environments and USP <797> applications.
These wipers are composed of exceptionally clean 100% meltblown polypropylene, pre-wetted with 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents.
Información relacionada
Certificados
Buscar por número de lote75831-354 data sheet AT-SatPax 550 SatPax550 Pre-wetted Meltblown Polypropylene Cleanroom Wiper SatPax® 550 TDS SatPax®550 Pre-wetted 100% Meltblown Polypropylene Cleanroom Wiper
Hoja de datos
VWR GHS Compliance Letter (US)
Detalles adicionales del producto
Frequently Bought Together