Extracta DBS, Quantabio
Extracta DBS is a rapid and easy to use extractions reagent optimized for the extraction of DNA from dried blood spot punches
- Optimized for DNA extraction from dried blood spot punches
- Single reagent for PCR-ready DNA in less than 30 minutes
- Maximized assay sensitivity, lower Ct’s, when combined with Quantabio ToughMix
- Compatible with high-throughput automation for PCR, qPCR and NGS applications
- Reliable, Consistent Lot-to-Lot Performance
Extracta DBS is a ready-to-use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman 903 filter paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing (1-3) reagents. Application of Extracta DBS with PerfeCta® qPCR ToughMix® or PerfeCta® MultiPlex qPCR ToughMix enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.
The resulting extract is immediately usable for downstream applications such as real-time PCR.
Extracta DBS is a rapid and easy to use extractions reagent optimized for the extraction of DNA from dried blood spot punches
- Optimized for DNA extraction from dried blood spot punches
- Single reagent for PCR-ready DNA in less than 30 minutes
- Maximized assay sensitivity, lower Ct’s, when combined with Quantabio ToughMix
- Compatible with high-throughput automation for PCR, qPCR and NGS applications
- Reliable, Consistent Lot-to-Lot Performance
Extracta DBS is a ready-to-use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman 903 filter paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing (1-3) reagents. Application of Extracta DBS with PerfeCta® qPCR ToughMix® or PerfeCta® MultiPlex qPCR ToughMix enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.
The resulting extract is immediately usable for downstream applications such as real-time PCR.
- Catalog No:
- 75877-508
- 75877-506
- Description:
- Extracta DBS PCR-ready genomic DNA from dried blood spots
- Extracta DBS PCR-ready genomic DNA from dried blood spots
- Target molecule:
- DNA
- DNA
- Refine target molecule:
- Total DNA
- Total DNA
- Sample type:
- Blood (dried) on filter cards
- Blood (dried) on filter cards
- Method / Format:
- Extraction for direct PCR
- Extraction for direct PCR
- Prep size:
- Scalable
- Scalable
Product Family Options
- DescriptionVolume
- Extracta DBS PCR-ready genomic DNA from dried blood spots10 mlEach (10ml)$1,621.41: 95171-010
Specifications
Cat. No.75877-508DescriptionExtracta DBS PCR-ready genomic DNA from dried blood spotsTarget moleculeDNARefine target moleculeTotal DNASample typeBlood (dried) on filter cardsMethod / FormatExtraction for direct PCRPrep sizeScalable - Extracta DBS PCR-ready genomic DNA from dried blood spots500 mlEach (500ml)$20,852.04: 95171-500
Specifications
Cat. No.75877-506DescriptionExtracta DBS PCR-ready genomic DNA from dried blood spotsTarget moleculeDNARefine target moleculeTotal DNASample typeBlood (dried) on filter cardsMethod / FormatExtraction for direct PCRPrep sizeScalable