Orders LinkContactUsLinkComponent
Syringe Extender, S17 Science
Syringe Extender, S17 Science
Syringe Extender with Luer Lock
Catalog # 470102-726
Supplier:   S17 SCIENCE LLC.
Número de proveedor #:  EQ 077
Syringe Extender, S17 Science
Catalog # 470102-726
Supplier:  S17 SCIENCE LLC.
Syringe Extender, S17 Science
Syringe Extender, S17 Science
Syringe Extender with Luer Lock
Price
$74.64
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
The extender is used to make an air-tight connection between a syringe and plastic, vinyl, or rubber tubing.
Especificaciones
  • Description:
    Syringe Extender with Luer Lock