VWR® Crystallizing Dishes
Supplier: VWR International
Jump to:
Acerca de este artículo
Made from high quality Type 1 Class A, 33 expansion Borosilicate glass, these crystallizing dishes have straight sides and flat bottom.
10 Options Available Below
Product Details & Documents
Made from high quality Type 1 Class A, 33 expansion Borosilicate glass, these crystallizing dishes have straight sides and flat bottom.
Product Family Options
Información del producto
- CapacityØHeightAvailabilityPrice
- 50 ml50 mm35 mmPaquete de 6$1,561.29Case de 4 (6UN)$1,701.81
- 80 ml60 mm35 mmPaquete de 6$1,548.93Case de 4 (6UN)$1,688.34
- 160 ml70 mm50 mmPaquete de 6$1,937.34Case de 4 (6UN)$2,111.71
- 270 ml90 mm50 mmPaquete de 6$1,722.53Case de 3 (6UN)$1,877.55
- 170 ml80 mm40 mmPaquete de 6$1,634.91Case de 4 (6UN)$1,782.05
- 340 ml100 mm50 mmPaquete de 6$1,938.57Case de 3 (6UN)$2,113.04
- 700 ml125 mm65 mmPaquete de 4$1,464.00Case de 3 (4UN)$1,595.76
- 1200 ml150 mm75 mmPaquete de 4$1,896.12Case de 2 (4UN)$2,066.77
- 1800 ml170 mm90 mmPaquete de 4$2,462.59Case de 2 (4UN)$2,684.23
- 2600 ml190 mm100 mmPaquete de 3$1,943.43Case de 2 (3UN)$2,118.33