LaboPlast®/SteriPlast® DispoLadle, Bürkle
Catalog # 75876-062
Supplier: BURKLE INC
Specifications
- Esterilidad:Sterile
- Descripción:DispoLadle
- Longitud:500 mm (19.69")
- Referencia:75876-062
- Nº de proveedor:5393-6641
Specifications
About this item
DispoLadle is a sampling scoop specially designed for disposable use
- 200 mL (6.76 oz.) scoop
- External diameter of 92 mm (3.62")
- Ergonomic handle
- HDPE construction
- Cleanroom manufactured, individually packaged
- Optionally sterilized by gamma rays
The scaled scoop with embossing can be used for measuring, and displays contents in increments of 10mL (0.34oz.) up to 200mL (6.76oz.). An ergonomic handle and a spout on both sides for right- or left-handed operation make handling simple and easy.
Packaging: 20 DispoLadles per package.