SP Bel-Art H-B Frio-Temp Calibrated Electronic Lollipop Thermometers for Refrigerators, Incubators and General Applications, Bel-Art Products, a part of SP
Catalog # 75854-038
Supplier: Bel-Art Products, a Part of SP
Specifications
- Resolución:0.1˚C/˚F
- Tipo de batería:CR2032
- Referencia:75854-038
- An x Al:38×114 mm (1.5×4.5")
- Puntos de calibración:37 °C Incubator
- Exactitud:±0.5 °C
- Sonda:50 mm (2") stainless steel probe in 30 ml plastic bottle/glass beads
- Rango:0...70 ˚C (32...158 ˚F)
- Nº de proveedor:B60210-1500
Specifications
About this item
Digital verification thermometers verify sample temperatures inside refrigerators and incubators without the use of a cable.
- Accurate temperature reading without sample contamination is achieved by using the liquid or glass bead filled bottle (included), which mimics the sample
- All models include accredited calibration documentation that meets the requirements of ISO 17025
- Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
- Individually serialized, CE marked
- Max./Min.
Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Certifications: Meets the requirements of ISO 17025.
Caution: Note: Not for use in ovens or freezers.