Orders LinkContactUsLinkComponent
Ice Borer, Bürkle
Ice borer complete, 50 mm (1.97") ID, 200 mm (7.87"), 54 mm (2.13")
  75806-596
 :   BURKLE INC
 :  5323-2010
Ice Borer, Bürkle
  75806-596
Ice Borer, Bürkle
Ice Borer, Bürkle
Ice borer complete, 50 mm (1.97") ID, 200 mm (7.87"), 54 mm (2.13")
$28,962.84
Each (1UN)
Sampler for deep-frozen and semi-solid substances products in the food industry.
  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilizable
  • Complete with case
The sharp sawing crown of the hollow borer rapidly cuts into the sample
It consists of three parts: borer, borer head for use in conventional drills and rod for pressing the cylindrical sample out of the borer head.
A cylindrical sample (cross sectional sample) with a diameter of 1.6 cm and a length of up to 20 cm can be cut out.
  • Ø:
    50 mm (1.97") ID
  • Ext.Ø:
    54 mm (2.13")
  • Description:
    Ice borer complete
  • Insertion Depth:
    200 mm (7.87")