Specifications
- Resolución:0.01°C
- Probe cable length:10' (3 m)
- W×D×H:23/4 × 41/4 × 3/4"
- Peso:142 g (5 oz.)
- Referencia:10860-788
- Exactitud:±0.25 °C
- Sonda:1 bottle probe (glycol) and 1 Bullet™ probe
- Rango:–50.00…+70.00 °C (–58.00…+158.00 °F)
Specifications
About this item
Datalogging thermometer that can be used in 21 CFR 11 environments.
- Features: MIN/MAX and current temperatures, Smart-Alarm(TM), °F/°C switchable
- Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
- Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
- Datalogging thermometers with user-defined time intervals
- Capable of storing over 1 million temperature observations and 10 unique alarm events
- Recorded data (CSV file) may be transferred from thermometer to PC or Mac using a USB flash drive - no additional software needed
- Status indicators - low battery, low memory, USB data transfer, active alarm state
Hi/Lo Alarms and Time/Date Stamps–Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments. Data for up to 10 different alarm events can be retrieved with the most current event viewable on the display. Smart-Alarm™ signals out-of-range conditions–When an out-of-range condition is sensed, the Smart-Alarm™ triggers. Smart-Alarm™ features visual and audible indicators which signal continuously until acknowledged, even if the parameter returns to the non-alarm range.
USB data transferring–All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac) using a USB flash drive (not included). No software is needed for computer interface. Each thermometer has a unique ID allowing multiple units to be used in the same location. Raw data on unit cannot be overwritten or modified. Indicator light will appear when 95% of memory is full.
Data-Logging Features–Can be used in environments requiring 21 CFR 11 compliance. Thermometer logs 2 years of observations with a 1 minute logging interval. Device will stop recording when storage is full and will not overwrite recorded data. Data is unable to be deleted from memory once stored.
Bottle probe features–Bottle probes are sealed in a miniature bottle (1 x 2-1/2 inches) filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about incidental contact with food or drinking water. Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened. Ideal for use in monitoring storage conditions for vaccines or other important samples. Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program.
Thermometer is supplied with batteries, 10 foot cable, magnetic strips, Velcro® material, bench stand, Traceable® Certificate. Unit comes with 4 extra batteries and a full 2-years of time on the calibration certificate.