Science Take-Out® Lung Disease
Proveedor: SCIENCE TAKE-OUT, LLC
Certificados
Acerca de este artículo
This complete experiment setup contains all materials and does not require lab equipment.
- Available as a Single Kit with Material for One Student or Group
- Unassembled 10 Pack for Additional Savings
- No Additional Equipment Needed
Students follow the case of a mother with chronic obstructive pulmonary disease (COPD) and conduct simulated genetic tests to determine if she and her son have Alpha-1 deficiency, a genetic predisposition for COPD.
They then model the correlation between COPD and Alpha-1 deficiency and explore environmental and occupational factors that increase the risk for COPD.
Frequently Bought Together