SP Wilmad-LabGlass 90° Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
14202-808
- ST Joint:14/20
- Referencia:14202-808
- Hose Connection O.D.:⁵⁄₁₆"
- Nº de proveedor:ML-195-700
Featuring a 90° bend to hose connector at top and a Standard Taper inner joint at bottom.
- 90° angle bend
- Top hose connector
- Bottom inner [ST] joint
Adapters are made of borosilicate glass.