Difco™ Glycerol, BD
Catalog # 90001-978
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Descripción:Glycerol
- Tamaño:500 g
- Referencia:90001-978
- Nº de proveedor:228220
Specifications
About this item
Highly purified alcohol used as a fixative, in bacterial preservation media and in the isolation and cultivation of many organisms.