Orders LinkContactUsLinkComponent
Difco™ EC Medium, BD Diagnostics
Difco™ EC Medium, BD Diagnostics
Certificates
Difco™ EC Medium, BD Diagnostics
2 Options Available Below
Product Details & Documents
Differentiation and enumeration of fecal and nonfecal coliforms in water, wastewater, shellfish and foods.
Product Family Options
Información del producto
  • Size
    Availability
    Price
  • EC Medium
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $5,370.00
    $5,370.00
    Each (500g)
       $5,370.00
    Supplier Number: 231430
  • 90001-606
    EC Medium with MUG
    EC Medium with MUG
    500 g
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (500g)
    Each (500g) - $18,840.00
    $18,840.00
    Each (500g)
       $18,840.00
    Supplier Number: 222200