Orders LinkContactUsLinkComponent
Difco™ Fraser Broth, BD Diagnostics
Difco™ Fraser Broth, BD Diagnostics
BD Difco™ fraser broth base, 500 g
Catalog # 90000-342
Supplier #:  211767
Difco™ Fraser Broth, BD Diagnostics
Catalog # 90000-342
CAS Number:  
Difco™ Fraser Broth, BD Diagnostics
BD Difco™ fraser broth base, 500 g
CAS Number:  
Price
$5,580.00
Log in Para disponibilidad
Each (500g)
Product Details & Documents
Selective enrichment and detection of Listeria.
Documents
Especificaciones
Especificaciones
  • Description:
    BD Difco™ fraser broth base
  • Size:
    500 g