AquaSampler, Bürkle
AquaSampler; 40 cm (15.75") Total Length, 46 mm (1.81"), 350 ml (11.83 oz.)
Catalog # 75846-218
Supplier: BURKLE INC
Número de proveedor #: 5308-0350
Jump to:
Product Details & Documents
Sampler for ground water samples from wells, pits, and other sources
- FEP/PTFE ultra pure, anti-adhesive surface
- Supplied without cable
Fits all normal well shafts with control openings to a diameter of 2", without tilting or getting stuck. Ultra pure materials guarantee the highest degree of sample purity. Easy to clean thanks to rounded edges and threads.
Packaging: Supplied one sampler per package.
Especificaciones
- Ø:46 mm (1.81")
- Capacity:350 ml (11.83 oz.)
- Description:AquaSampler; 40 cm (15.75") Total Length