ContactUsLinkComponent
Bacto M Broth, BD Diagnostics
  90004-538
 :  
Bacto M Broth, BD Diagnostics
  90004-538
 :  294020
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Descripción:
    M Broth
  • Tamaño:
    500 g
  • Referencia:
    90004-538
  • Nº de proveedor:
    294020
Bacto™ M broth is used for cultivating Salmonella in foods and feeds by the accelerated enrichment serology (ES) procedure.
  • Light amber, clear to very slightly opalescent, may have a slight precipitate
  • pH 7.0 ±0.2
Thsi broth is prepared according to the formula of Sperber and Diebel and contains all the nutrients necessary for good growth
and flagella development of Salmonella.
Thsi broth also conforms to the testing standards recommended by the Compendium of Methods for the Microbiological
Examination of Foods4 (APHA).