Full Color Polarizing Polariscope with Swivel Mount, Ace Glass
75846-508
- Descripción:Full Color Polariscope with 10x19" Illuminated Area and 5" Square Viewer
- Referencia:75846-508
- Detalles eléctricos:120 V, 60 Hz
- Nº de proveedor:14400-10
Full color polarizer cabinet for viewing strain patterns in glass and other transparent materials.
- Swivel mount allows cabinet to be locked into any horizontal or vertical position
Polariscope consists of a 4" deep cabinet with a 10x19" illuminated area, and a 5" square viewer.
: One-year manufacturer’s warranty.