Orders LinkContactUsLinkComponent
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Certificates
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
2 Options Available Below
Product Details & Documents
Difco™ Fluid Lactose Medium is used in microbiology for the cultivation of Salmonella and coliform organisms in water, food, dairy and pharmaceutical products.
  • Salmonella cultivation
  • Coliform organism cultivation
Documents
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 90000-392
    Difco™ Fluid lactose medium
    Difco™ Fluid lactose medium
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $3,110.00
    $3,110.00
    Each (500g)
       $3,110.00
    Supplier Number: 211835
  • 90000-910
    Difco™ Lactose monohydrate
    Difco™ Lactose monohydrate
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $2,520.00
    $2,520.00
    Each (500g)
       $2,520.00
    Supplier Number: 215620